Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153582.5 | NP_705810.2 | 4 | 727 | ||
GenBank mRNA | AF479815.1 | - | 4 | 616 | ||
BC028852.1 | - | 4 | 572 | 4206772 | ||
BE952164.1 | - | 4 | 616 |