Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172054.4 | NP_742051.1 | 5 | 1277 | ||
GenBank mRNA | AK002893.1 | - | 5 | 1282 | ||
AK028525.1 | - | 5 | 1274 | |||
AK153857.1 | - | 5 | 1278 |