Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136528.1 | NP_001130000.1 | 9 | 1654 | ||
NM_001136530.1 | NP_001130002.1 | 9 | 1584 | |||
NM_006216.3 | NP_006207.1 | 9 | 1657 | |||
NR_073116.1 | 10 | 2072 | ||||
XM_005246641.2 | XP_005246698.1 | 9 | 1746 | |||
XM_017004329.1 | XP_016859818.1 | 9 | 1638 | |||
XM_017004330.1 | XP_016859819.1 | 9 | 1475 | |||
XM_017004331.1 | XP_016859820.1 | 9 | 1635 | |||
XM_017004332.1 | XP_016859821.1 | 9 | 1472 | |||
GenBank mRNA | AJ293400.1 | - | 1 | 34 | ||
AK225823.1 | - | 9 | 1622 | |||
BC015663.2 | - | 9 | 1613 | 4578406 | ||
BC042628.1 | - | 10 | 2071 | 4824856 | ||
BX360097.2 | - | 1 | 227 | |||
BX647459.1 | - | 2 | 5489 |