Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012303.2 | NP_001012303.1 | 5 | 798 | ||
NM_006217.4 | NP_006208.1 | 4 | 707 | |||
GenBank mRNA | AB006423.1 | - | 4 | 624 | ||
AF130470.1 | - | 3 | 633 | |||
AK226025.1 | - | 4 | 597 | |||
AK293391.1 | - | 4 | 623 | |||
BC027859.2 | - | 4 | 668 | 5228038 | ||
BX496933.1 | - | 4 | 624 |