Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175247.3 | NP_780456.2 | 7 | 3036 | ||
GenBank mRNA | AK013265.1 | - | 1 | 409 | ||
AK048530.1 | - | 7 | 3028 | |||
AK173170.1 | - | 7 | 3001 |