Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006224.3 | NP_006215.1 | 11 | 1057 | ||
XM_011523939.1 | XP_011522241.1 | 10 | 969 | |||
XM_011523940.1 | XP_011522242.1 | 11 | 1479 | |||
GenBank mRNA | AK303986.1 | - | 11 | 827 | ||
BC027177.1 | - | 9 | 904 | |||
BC045108.1 | - | 11 | 1032 | 5454273 | ||
BC068538.1 | - | 11 | 940 | |||
BC082976.1 | - | 11 | 972 | 6578454 | ||
D30036.1 | - | 11 | 1009 | |||
EU794647.1 | - | 11 | 1032 | |||
M73704.1 | - | 11 | 1017 |