Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166531.1 | NP_001160003.1 | 22 | 7024 | ||
NM_001166532.1 | NP_001160004.1 | 21 | 7118 | |||
NM_019460.2 | NP_062333.1 | 21 | 6975 | |||
GenBank mRNA | AK020390.1 | - | 383 | |||
AK029771.1 | - | 2958 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_175254.1 | - | 383 | Chr. - - [-] on Build |