Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNLIPRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303135.1 | NP_001290064.1 | 11 | 1178 | ||
NM_006229.3 | NP_006220.1 | 11 | 1188 | |||
XM_011539868.1 | XP_011538170.1 | 11 | 1321 | |||
XM_011539869.1 | XP_011538171.1 | 11 | 1589 | |||
GenBank mRNA | AK130236.1 | - | 11 | 1176 | ||
BC025784.2 | - | 10 | 1151 | 5226231 | ||
CR749299.1 | - | 10 | 2947 | |||
CR749524.1 | - | 10 | 3103 | |||
CR749596.1 | - | 10 | 3101 | |||
M93283.1 | - | 11 | 1170 |