Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175286.4 | NP_780495.2 | 4 | 2675 | ||
GenBank mRNA | AK082888.1 | - | 4 | 2656 | ||
AK171799.1 | - | 4 | 2686 | |||
BC024573.1 | - | 4 | 980 | |||
BC047927.1 | - | 3 | 697 | |||
BC055478.1 | - | 4 | 1120 | |||
BC089508.1 | - | 4 | 2185 | 6589951 |