Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nucks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145804.1 | NP_001139276.1 | 5 | 570 | ||
NM_175294.3 | NP_780503.2 | 5 | 570 | |||
GenBank mRNA | AB049825.1 | - | 5 | 549 | ||
AK078816.1 | - | 5 | 494 | |||
BC039951.1 | - | 5 | 482 | 4188558 |