Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175309.4 | NP_780518.1 | 2 | 500 | ||
GenBank mRNA | AK042067.1 | - | 2 | 296 | ||
AK052525.1 | - | 2 | 502 | |||
AK157647.1 | - | 2 | 307 | |||
AY233463.1 | - | 2 | 294 | |||
BC061224.1 | - | 2 | 297 | |||
BC120594.1 | - | 2 | 250 | 40129517 | ||
BC137742.1 | - | 2 | 250 | 8860761 |