Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175309.4 | NP_780518.1 | 3 | 617 | ||
GenBank mRNA | AK042067.1 | - | 3 | 413 | ||
AK052525.1 | - | 3 | 619 | |||
AK157647.1 | - | 3 | 424 | |||
AY233463.1 | - | 3 | 411 | |||
BC061224.1 | - | 3 | 414 | |||
BC120594.1 | - | 3 | 367 | 40129517 | ||
BC137742.1 | - | 3 | 367 | 8860761 |