Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159672.1 | NP_001153144.1 | 5 | 375 | ||
NM_175317.3 | NP_780526.2 | 5 | 404 | |||
GenBank mRNA | AK028676.1 | - | 5 | 377 | ||
AK031647.1 | - | 5 | 361 | |||
AK049698.1 | - | 5 | 362 | |||
BC045616.1 | - | 5 | 358 | 4983234 |