Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175324.3 | NP_780533.2 | 13 | 1604 | ||
GenBank mRNA | AK077531.1 | - | 13 | 1605 | ||
AK128998.1 | - | 13 | 1583 | |||
AK139918.1 | - | 13 | 1548 | |||
AK145561.1 | - | 13 | 1593 | |||
AK145834.1 | - | 13 | 1584 | |||
BC006042.1 | - | 3 | 280 | |||
BC032994.1 | - | 6 | 533 | |||
BC045199.1 | - | 13 | 1568 | 5133197 |