Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175333.3 | NP_780542.1 | 7 | 982 | ||
GenBank mRNA | AK077111.1 | - | 7 | 983 | ||
BC115588.1 | - | 8 | 1081 | 40102382 | ||
BC115589.1 | - | 7 | 975 | 40102384 | ||
FM165286.1 | - | 7 | 924 |