Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113471.1 | NP_001106942.1 | 8 | 1242 | ||
NM_175640.2 | NP_783571.2 | 8 | 1298 | |||
GenBank mRNA | AK031445.1 | - | 7 | 1223 | ||
AY161165.1 | - | 7 | 1193 | |||
BC037748.1 | - | 1 | 16 | |||
BC096685.1 | - | 8 | 1243 | 1448913 |