Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175681.3 | NP_783612.2 | 2 | 409 | ||
GenBank mRNA | AY255567.1 | - | 1 | 23 | ||
AY605231.1 | - | 2 | 128 | |||
BC044746.2 | - | 2 | 409 | 5363416 |
Set Membership: |
G-protein coupled receptor |