Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006273.3 | NP_006264.2 | 3 | 335 | ||
GenBank mRNA | AF043338.1 | - | 2 | 191 | ||
BC070240.1 | - | 2 | 209 | |||
BC092436.1 | - | 3 | 329 | 30418073 | ||
BC112258.1 | - | 3 | 325 | 8327726 | ||
BC112260.1 | - | 3 | 325 | 8327728 | ||
CD523198.1 | - | 3 | 338 | |||
X71087.1 | - | 3 | 330 |