Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178405.3 | NP_848492.1 | 11 | 1596 | ||
GenBank mRNA | AK052863.1 | - | 11 | 1533 | ||
AK137602.1 | - | 11 | 1532 | |||
AK147331.1 | - | 11 | 1528 | |||
AK148775.1 | - | 11 | 1528 | |||
AK161840.1 | - | 11 | 1528 | |||
AK164509.1 | - | 11 | 1532 | |||
AK168703.1 | - | 11 | 1528 | |||
BC036127.1 | - | 11 | 1520 | 4955003 | ||
BC041774.1 | - | 8 | 1246 |