Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242349.1 | NP_001229278.1 | 25 | 2985 | ||
NM_178642.5 | NP_848757.4 | 26 | 2994 | |||
GenBank mRNA | AK028991.1 | - | 8 | 896 | ||
AK029329.1 | - | 12 | 1361 | |||
AK032793.1 | - | 21 | 2006 | |||
AK052589.1 | - | 26 | 2946 | |||
BC004697.1 | - | 18 | 1859 | |||
BC006062.1 | - | 9 | 944 | |||
BC062959.1 | - | 25 | 2871 | 30547439 |