Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271915.1 | NP_001258844.1 | 1 | 1802 | ||
NM_178879.4 | NP_849210.1 | 2 | 1614 | |||
GenBank mRNA | AK032108.1 | - | 1 | 1803 | ||
AK053867.1 | - | 1 | 1316 | |||
AK169360.1 | - | 1 | 1798 | |||
BC047209.1 | - | 1 | 1774 | |||
BC108347.1 | - | 1 | 1752 | 5010813 | ||
BC120583.1 | - | 1 | 1467 | 40129506 | ||
BC120585.1 | - | 1 | 1467 | 40129508 |