Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178883.5 | NP_849214.2 | 2 | 183 | ||
GenBank mRNA | AK039170.1 | - | 2 | 190 | ||
AK043953.1 | - | 2 | 184 | |||
AK165909.1 | - | 2 | 153 | |||
AK169151.1 | - | 2 | 176 | |||
AK170455.1 | - | 2 | 200 | |||
BC119132.1 | - | 2 | 166 | 8733881 | ||
BC119134.1 | - | 2 | 166 | 8733883 |