Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178883.5 | NP_849214.2 | 5 | 2032 | ||
GenBank mRNA | AK043953.1 | - | 5 | 2033 | ||
AK165909.1 | - | 5 | 2002 | |||
AK169151.1 | - | 5 | 2025 | |||
AK170455.1 | - | 6 | 2191 | |||
BC013244.1 | - | 2 | 1416 | |||
BC013262.1 | - | 2 | 1416 |