Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005246818.1 | XP_005246875.1 | 3 | 427 | ||
XM_005246819.1 | XP_005246876.1 | 2 | 175 | |||
XM_006712720.3 | XP_006712783.1 | 3 | 214 | |||
XM_011511707.2 | XP_011510009.1 | 4 | 555 | |||
XM_011511708.1 | XP_011510010.1 | 3 | 489 | |||
XM_011511709.2 | XP_011510011.1 | 3 | 257 | |||
GenBank mRNA | AB209866.1 | - | 2 | 6349 | ||
AF021834.1 | - | 3 | 371 | |||
AK313562.1 | - | 2 | 339 | |||
AU136083.1 | - | 3 | 518 | |||
BC015514.1 | - | 2 | 341 | 3902987 | ||
DB078397.1 | - | 2 | 397 | |||
HY175080.1 | - | 2 | 410 | |||
J03225.1 | - | 2 | 233 |