Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178884.5 | NP_849215.3 | 4 | 1987 | ||
GenBank mRNA | AK029863.1 | - | 4 | 1777 | ||
AK031074.1 | - | 4 | 1988 | |||
AK034852.1 | - | 3 | 2049 | |||
AK048144.1 | - | 4 | 1179 | |||
AK162473.1 | - | 4 | 1897 | |||
AK202760.1 | - | 1 | 26 | |||
BC051643.1 | - | 2 | 539 | |||
BC056205.1 | - | 1 | 161 |