Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178890.3 | NP_849221.2 | 6 | 2061 | ||
GenBank mRNA | AK042322.1 | - | 6 | 2060 | ||
AK151620.1 | - | 6 | 2057 | |||
AK152163.1 | - | 6 | 2057 | |||
AK170569.1 | - | 6 | 2060 | |||
AK172414.1 | - | 6 | 2059 | |||
AK172468.1 | - | 6 | 2058 | |||
AK182096.1 | - | 2 | 110 | |||
BC054399.1 | - | 6 | 2061 | 6493840 |