Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178890.3 | NP_849221.2 | 14 | 3082 | ||
GenBank mRNA | AK042322.1 | - | 14 | 3082 | ||
AK151620.1 | - | 14 | 3078 | |||
AK152163.1 | - | 14 | 3078 | |||
AK170569.1 | - | 14 | 3081 | |||
AK172414.1 | - | 14 | 3080 | |||
AK172468.1 | - | 14 | 3079 | |||
BC006756.1 | - | 5 | 622 | |||
BC054399.1 | - | 14 | 3082 | 6493840 |