Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190733.1 | NP_001177662.1 | 10 | 1217 | ||
NM_001190734.1 | NP_001177663.1 | 12 | 1257 | |||
NM_178896.5 | NP_849227.1 | 11 | 1322 | |||
GenBank mRNA | AK033559.1 | - | 11 | 1267 | ||
AK081894.1 | - | 1 | 114 | |||
BC031793.1 | - | 2 | 446 | |||
BC060213.1 | - | 11 | 1264 | |||
BC108346.1 | - | 11 | 1224 | 6310392 | ||
BC137627.1 | - | 11 | 1266 | 8860646 | ||
BC137628.1 | - | 11 | 1266 | 8860647 | ||
BC144794.1 | - | 10 | 1161 | 9053342 |