Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178907.3 | NP_849238.1 | 11 | 1941 | ||
GenBank mRNA | AK051962.1 | - | 9 | 2863 | ||
AK087496.1 | - | 10 | 1603 | |||
AK144637.1 | - | 11 | 1879 | |||
AK151881.1 | - | 11 | 1864 | |||
AK170456.1 | - | 11 | 1937 | |||
AK172344.1 | - | 11 | 1853 | |||
BC031467.1 | - | 11 | 1942 | 4189474 |