Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181397.2 | NP_852062.1 | 10 | 2482 | ||
GenBank mRNA | AK009372.1 | - | 1 | 705 | ||
AK082517.1 | - | 2095 | ||||
AK087803.1 | - | 2256 | ||||
AK088466.1 | - | 8 | 2019 | |||
AK152130.1 | - | 10 | 2404 | |||
AK153288.1 | - | 10 | 2404 | |||
AK155870.1 | - | 10 | 2446 | |||
AK172884.1 | - | 10 | 2410 | |||
BC043324.1 | - | 6 | 1532 | |||
BC052074.1 | - | 10 | 2482 | 6401192 |