Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146158.1 | NP_001139630.1 | 3 | 345 | ||
NM_006302.2 | NP_006293.2 | 2 | 671 | |||
GenBank mRNA | AK292553.1 | - | 2 | 617 | ||
AK307991.1 | - | 3 | 331 | |||
AK313355.1 | - | 2 | 671 | |||
BC028337.1 | - | 2 | 652 | 5110525 | ||
BC117250.1 | - | 2 | 544 | 40125801 | ||
BC117252.1 | - | 2 | 544 | 40125803 | ||
DC388420.1 | - | 3 | 317 | |||
X87237.1 | - | 2 | 640 |