Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181401.3 | NP_852066.2 | 3 | 2560 | ||
GenBank mRNA | AK035894.1 | - | 3 | 2247 | ||
AK156282.1 | - | 3 | 2605 | |||
AK162736.1 | - | 1 | 642 | |||
AK169861.1 | - | 3 | 2217 | |||
AK170682.1 | - | 3 | 2273 | |||
AY147882.1 | - | 3 | 2559 | |||
BC030341.1 | - | 3 | 2564 | 3661112 |