Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Champ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181854.2 | NP_862902.1 | 2 | 441 | ||
GenBank mRNA | AK129444.1 | - | 2 | 341 | ||
AK132164.1 | - | 2 | 404 | |||
AK133990.1 | - | 2 | 367 | |||
AK139963.1 | - | 2 | 406 | |||
AK170835.1 | - | 2 | 384 | |||
BC028991.1 | - | 2 | 353 | 5355869 |