Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194336.2 | NP_919317.2 | 11 | 3119 | ||
GenBank mRNA | AK128993.1 | - | 3114 | |||
AK129021.1 | - | 2390 | ||||
BC007143.1 | - | 10 | 2975 | 3495728 | ||
BC115768.1 | - | 3114 | 40085916 |