Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198163.1 | NP_937806.1 | 6 | 1630 | ||
GenBank mRNA | AK020563.1 | - | 1 | 351 | ||
AK156576.1 | - | 6 | 1621 | |||
BC030941.1 | - | 4 | 1321 | |||
BC056466.1 | - | 6 | 1630 | 5717246 |