Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198190.1 | NP_937833.1 | 3 | 1140 | ||
GenBank mRNA | AK019330.1 | - | 1 | 57 | ||
BC052191.1 | - | 3 | 1140 | 6505258 |