Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023579.4 | NP_076068.1 | 4 | 449 | ||
GenBank mRNA | AF294327.1 | - | 4 | 400 | ||
AK017701.1 | - | 4 | 408 | |||
AK049422.1 | - | 4 | 421 | |||
AK053681.1 | - | 4 | 414 | |||
AK079134.1 | - | 4 | 419 | |||
AK167303.1 | - | 4 | 411 | |||
AK169664.1 | - | 4 | 417 | |||
BC052392.1 | - | 4 | 392 | 5719784 | ||
BC054814.1 | - | 4 | 622 | 6842146 | ||
CB524964.1 | - | 4 | 408 |