Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023579.4 | NP_076068.1 | 5 | 591 | ||
GenBank mRNA | AF294327.1 | - | 5 | 542 | ||
AK017701.1 | - | 5 | 550 | |||
AK049422.1 | - | 5 | 563 | |||
AK053681.1 | - | 5 | 556 | |||
AK079134.1 | - | 5 | 561 | |||
AK167303.1 | - | 5 | 553 | |||
AK169664.1 | - | 5 | 559 | |||
BC052392.1 | - | 5 | 534 | 5719784 | ||
BC054814.1 | - | 5 | 764 | 6842146 | ||
CB524964.1 | - | 5 | 550 |