Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023585.4 | NP_076074.2 | 3 | 277 | ||
GenBank mRNA | AK019486.1 | - | 3 | 248 | ||
AK028472.1 | - | 3 | 251 | |||
AK028742.1 | - | 3 | 261 | |||
AK031233.1 | - | 3 | 246 | |||
AK033016.1 | - | 3 | 262 | |||
AK146073.1 | - | 3 | 249 | |||
BC029742.1 | - | 3 | 245 | |||
BC058374.1 | - | 3 | 248 | 5715168 | ||
BC083098.1 | - | 3 | 262 | 30460946 | ||
BC094903.1 | - | 3 | 240 | 30081846 | ||
BY025619.1 | - | 3 | 285 |