Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 3 | 369 | ||
NM_001252079.1 | NP_001239008.1 | 3 | 369 | |||
NM_006313.2 | NP_006304.1 | 3 | 369 | |||
XM_006719718.3 | XP_006719781.1 | 3 | 367 | |||
XM_017020285.1 | XP_016875774.1 | 3 | 320 | |||
GenBank mRNA | AB011101.2 | - | 3 | 294 | ||
AF013990.1 | - | 3 | 240 | |||
AF106069.1 | - | 3 | 303 | |||
AF153604.1 | - | 3 | 351 | |||
AK023703.1 | - | 3 | 312 | |||
AK292337.1 | - | 3 | 329 | |||
AK295947.1 | - | 3 | 301 | |||
AK297759.1 | - | 3 | 305 | |||
BC020688.1 | - | 3 | 305 | |||
BC063454.1 | - | 3 | 309 | |||
BC110908.1 | - | 3 | 302 | |||
BC125123.1 | - | 3 | 303 | 40118994 | ||
DB451322.1 | - | 3 | 371 |