Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159351.1 | NP_001152823.1 | 3 | 343 | ||
NM_023585.4 | NP_076074.2 | 4 | 469 | |||
GenBank mRNA | AF303828.1 | - | 3 | 426 | ||
AK017786.1 | - | 3 | 324 | |||
AK019486.1 | - | 4 | 440 | |||
AK028472.1 | - | 4 | 443 | |||
AK028742.1 | - | 4 | 453 | |||
AK031233.1 | - | 4 | 438 | |||
AK033016.1 | - | 4 | 454 | |||
AK146073.1 | - | 4 | 441 | |||
BC029742.1 | - | 4 | 437 | |||
BC044722.1 | - | 3 | 307 | |||
BC058374.1 | - | 4 | 440 | 5715168 | ||
BC083098.1 | - | 4 | 454 | 30460946 | ||
BC094903.1 | - | 4 | 432 | 30081846 | ||
CJ049747.1 | - | 3 | 327 |