Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023596.3 | NP_076085.1 | 6 | 1561 | ||
GenBank mRNA | AF326986.1 | - | 6 | 1572 | ||
AK034133.1 | - | 6 | 1557 | |||
AK051317.1 | - | 6 | 1586 | |||
AK157868.1 | - | 5 | 1399 | |||
BC137864.1 | - | 6 | 1544 | 8860883 | ||
BC144983.1 | - | 6 | 1545 |