Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023596.3 | NP_076085.1 | 6 | 2186 | ||
GenBank mRNA | AK034133.1 | - | 6 | 2183 | ||
AK157868.1 | - | 5 | 2024 | |||
BC137864.1 | - | 6 | 2169 | 8860883 | ||
BC144983.1 | - | 6 | 2170 |