Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081490.2 | NP_001074959.1 | 3, 4 | 315 | ||
NM_023605.2 | NP_076094.2 | 3, 4 | 617 | |||
GenBank mRNA | AB041585.1 | - | 3, 4 | 586 | ||
AK018482.1 | - | 3, 4 | 568 | |||
AK077607.1 | - | 3, 4 | 278 | |||
BC020074.1 | - | 3, 4 | 588 | 3710611 | ||
DV657928.1 | - | 3, 4 | 303 |