Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023625.4 | NP_076114.2 | 7, 8 | 1173 | ||
GenBank mRNA | AK004973.1 | - | 7, 8 | 1176 | ||
AK029813.1 | - | 7, 8 | 1173 | |||
AK030234.1 | - | 7, 8 | 1125 | |||
AK032930.1 | - | 7, 8 | 1169 | |||
AK042089.1 | - | 1169 | ||||
AK043608.1 | - | 7, 8 | 1163 | |||
AK046937.1 | - | 7, 8 | 1163 | |||
AK150201.1 | - | 7, 8 | 1162 | |||
AK164220.1 | - | 7, 8 | 1154 | |||
AK170387.1 | - | 7, 8 | 1165 | |||
AK170631.1 | - | 7, 8 | 1153 | |||
BC026395.1 | - | 7, 8 | 1151 | |||
BC038605.1 | - | 7, 8 | 1154 | 4921245 | ||
BC048826.1 | - | 7, 8 | 1160 | 4196604 | ||
BC060095.1 | - | 5, 6 | 2764 |