Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023626.4 | NP_076115.3 | 9 | 883 | ||
GenBank mRNA | AK030364.1 | - | 8 | 690 | ||
AK080787.1 | - | 9 | 852 | |||
AK131993.1 | - | 9 | 883 | |||
AK146145.1 | - | 8 | 666 | |||
AY007791.1 | - | 9 | 852 | |||
BC005721.1 | - | 2 | 72 |