Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317333.1 | NP_001304262.1 | 4 | 578 | ||
NM_006321.3 | NP_006312.1 | 3 | 456 | |||
XM_011533265.1 | XP_011531567.1 | 3 | 444 | |||
XM_011533266.1 | XP_011531568.1 | 4 | 566 | |||
XM_011533267.1 | XP_011531569.1 | 3 | 498 | |||
XM_011533268.1 | XP_011531570.1 | 2 | 391 | |||
XM_011533270.1 | XP_011531572.1 | 3 | 432 | |||
XM_011533273.1 | XP_011531575.1 | 3 | 432 | |||
XM_011533275.2 | XP_011531577.1 | 2 | 352 | |||
XM_017005531.1 | XP_016861020.1 | 3 | 498 | |||
XM_017005532.1 | XP_016861021.1 | 2 | 391 | |||
XM_017005533.1 | XP_016861022.1 | 1 | 792 | |||
XM_017005536.1 | XP_016861025.1 | 4 | 566 | |||
XM_017005537.1 | XP_016861026.1 | 3 | 432 | |||
XM_017005538.1 | XP_016861027.1 | 3 | 478 | |||
XM_017005540.1 | XP_016861029.1 | 3 | 432 | |||
XM_017005541.1 | XP_016861030.1 | 3 | 502 | |||
GenBank mRNA | AF099149.1 | - | 2 | 260 | ||
AF099150.1 | - | 2 | 260 | |||
AF099151.1 | - | 2 | 260 | |||
AF099152.1 | - | 2 | 260 | |||
AF183427.1 | - | 2 | 259 | |||
AJ130978.1 | - | 2 | 260 | |||
AK001800.1 | - | 1 | 140 | |||
AK091240.1 | - | 1 | 957 | |||
AK223550.1 | - | 4 | 562 | |||
AK313434.1 | - | 1 | 120 | |||
BC000422.2 | - | 3 | 416 | 2964505 | ||
CN399800.1 | - | 3 | 430 | |||
CR456785.1 | - | 1 | 116 |