Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085383.1 | NP_001078852.1 | 13 | 1804 | ||
NM_023628.2 | NP_076117.2 | 13 | 1783 | |||
GenBank mRNA | AJ401160.1 | - | 13 | 1737 | ||
AK010128.1 | - | 13 | 1784 | |||
BC062140.1 | - | 13 | 1794 | 30290956 |