Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023630.2 | NP_076119.2 | 7 | 1198 | ||
GenBank mRNA | AF250835.1 | - | 7 | 1166 | ||
AK005881.1 | - | 7 | 1173 | |||
BC050756.1 | - | 7 | 1181 | 6741860 |