Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023631.2 | NP_076120.2 | 14 | 1556 | ||
GenBank mRNA | AF233581.1 | - | 14 | 1483 | ||
AK158427.1 | - | 14 | 1557 | |||
AK160953.1 | - | 14 | 1522 | |||
BC117975.1 | - | 14 | 1483 | 40098789 | ||
BC117976.1 | - | 14 | 1483 | 40098791 |